ENA282548407 Enamine

(2-amino-6-bromophenyl)methanol

  • MDL number MFCD11036299
  • Cas Number : 861106-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60178

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60178
Availability details for
Quantity
  • Availability
  • Add to Cart