ENA282548419 Enamine

1-cyclopentyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09046556
  • Cas Number : 926262-55-7
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61034
Availability details for
Quantity
  • Availability
  • Add to Cart