ENA282548420 Enamine

bis(4-chlorophenyl)-1,2-oxazol-5-amine

  • MDL number MFCD12653035
  • Cas Number : 1096934-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60190
Availability details for
Quantity
  • Availability
  • Add to Cart