ENA282548426 Enamine

3-tert-butyl-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09045788
  • Cas Number : 926257-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61044
Availability details for
Quantity
  • Availability
  • Add to Cart