ENA282548432 Enamine

4-(3-cyanopropanesulfonyl)benzoic acid

  • MDL number MFCD09942888
  • Cas Number : 1016834-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61051
Availability details for
Quantity
  • Availability
  • Add to Cart