ENA282548433 Enamine

4-(4-chlorophenyl)-3-(4-fluorophenyl)-1,2-oxazol-5-amine

  • MDL number MFCD12541217
  • Cas Number : 1097153-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60200
Availability details for
Quantity
  • Availability
  • Add to Cart