ENA282548435 Enamine

3-[(3-methylphenyl)amino]propanamide

  • MDL number MFCD11205137
  • Cas Number : 1060675-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61055
Availability details for
Quantity
  • Availability
  • Add to Cart