ENA282548436 Enamine

2-[(4-sulfamoylphenyl)formamido]acetic acid

  • MDL number MFCD08445033
  • Cas Number : 143288-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61056
Availability details for
Quantity
  • Availability
  • Add to Cart