ENA282548440 Enamine

8-methoxy-2H-chromene-3-carboxylic acid

  • MDL number MFCD00202018
  • Cas Number : 57543-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50369
Availability details for
Quantity
  • Availability
  • Add to Cart