ENA282548442 Enamine

1-(2-hydroxy-5-phenylphenyl)ethan-1-one

  • MDL number MFCD06802890
  • Cas Number : 14031-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61064
Availability details for
Quantity
  • Availability
  • Add to Cart