ENA282548444 Enamine

2-[4-(cyclohexylamino)phenyl]acetonitrile

  • MDL number MFCD12038984
  • Cas Number : 1157685-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61066
Availability details for
Quantity
  • Availability
  • Add to Cart