ENA282548450 Enamine

2-(5-amino-1H-indol-1-yl)-1-(morpholin-4-yl)ethan-1-one

  • MDL number MFCD12654343
  • Cas Number : 1097161-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60222

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60222
Availability details for
Quantity
  • Availability
  • Add to Cart