ENA282548454 Enamine

5H,6H,7H,8H-[1,2,4]triazolo[3,4-a]pyridine-6-carboxylic acid

  • MDL number MFCD09701670
  • Cas Number : 1035841-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279...281
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51350

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51350
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart