ENA282548458 Enamine

3-(ethylamino)pyrrolidine-2,5-dione

  • MDL number MFCD12099318
  • Cas Number : 1218654-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61084
Availability details for
Quantity
  • Availability
  • Add to Cart