ENA282548464 Enamine

4-methyl-1-[(piperidin-3-yl)methyl]piperidine

  • MDL number MFCD09041230
  • Cas Number : 926212-17-1
  • Molecular Weight : 196.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61091
Availability details for
Quantity
  • Availability
  • Add to Cart