ENA282548465 Enamine

2-(2-ethoxyethoxy)benzoic acid

  • MDL number MFCD08134363
  • Cas Number : 367513-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61092

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61092
Availability details for
Quantity
  • Availability
  • Add to Cart