ENA282548475 Enamine

2-methyl-3-nitrobenzamide

  • MDL number MFCD00463807
  • Cas Number : 290366-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61115

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61115
Availability details for
Quantity
  • Availability
  • Add to Cart