ENA282548477 Enamine

3-[(4-aminophenyl)methyl]-1,3-thiazolidine-2,4-dione

  • MDL number MFCD09742330
  • Cas Number : 258856-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61117

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61117
Availability details for
Quantity
  • Availability
  • Add to Cart