ENA282548483 Enamine

2-(1-ethyl-1H-indol-3-yl)acetic acid

  • MDL number MFCD07401792
  • Cas Number : 58690-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61125
Availability details for
Quantity
  • Availability
  • Add to Cart