ENA282548486 Enamine

3-(furan-2-yl)propanehydrazide

  • MDL number MFCD08061556
  • Cas Number : 98334-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61129
Availability details for
Quantity
  • Availability
  • Add to Cart