ENA282548487 Enamine

1-(3-amino-2-methylphenyl)pyrrolidine-2,5-dione

  • MDL number MFCD09736080
  • Cas Number : 268738-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61130
Availability details for
Quantity
  • Availability
  • Add to Cart