ENA282548488 Enamine

1-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]-1H-pyrazol-3-amine

  • MDL number MFCD12603147
  • Cas Number : 1183703-75-4
  • Molecular Weight : 232.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60258

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60258
Availability details for
Quantity
  • Availability
  • Add to Cart