ENA282548491 Enamine

2-{[2-(4-chlorophenyl)-2-oxoethyl]sulfanyl}acetic acid

  • MDL number MFCD04067646
  • Cas Number : 153783-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61134

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61134
Availability details for
Quantity
  • Availability
  • Add to Cart