ENA282548495 Enamine

3-[3-(4H-1,2,4-triazol-3-yl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD11203663
  • Cas Number : 1038356-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61140
Availability details for
Quantity
  • Availability
  • Add to Cart