ENA282548498 Enamine

3-methyl-4-(1H-1,2,3,4-tetrazol-1-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD11650713
  • Cas Number : 1099632-63-9
  • Molecular Weight : 258.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60268
Availability details for
Quantity
  • Availability
  • Add to Cart