ENA282548509 Enamine

2-formylbenzene-1-sulfonyl chloride

  • MDL number MFCD10697733
  • Cas Number : 21639-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33...35
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61171
Availability details for
Quantity
  • Availability
  • Add to Cart