ENA282548511 Enamine

2-(4-amino-1H-pyrazol-1-yl)-N-(4-methoxyphenyl)acetamide

  • MDL number MFCD11128230
  • Cas Number : 1152853-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60288
Availability details for
Quantity
  • Availability
  • Add to Cart