ENA282548517 Enamine

ethyl 2-amino-5-phenylbenzoate

  • MDL number MFCD12602975
  • Cas Number : 889673-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61185
Availability details for
Quantity
  • Availability
  • Add to Cart