ENA282548522 Enamine

1-[4-(1H-1,2,4-triazol-1-yl)phenyl]ethan-1-ol

  • MDL number MFCD08442525
  • Cas Number : 926228-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61192
Availability details for
Quantity
  • Availability
  • Add to Cart