ENA282548523 Enamine

3-amino-1-[4-(pyridin-2-yl)piperazin-1-yl]propan-1-one

  • MDL number MFCD09726250
  • Cas Number : 938517-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61193
Availability details for
Quantity
  • Availability
  • Add to Cart