ENA282548524 Enamine

1,3-dimethyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-sulfonyl chloride

  • MDL number MFCD05237674
  • Cas Number : 24134-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-180
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60294
Availability details for
Quantity
  • Availability
  • Add to Cart