ENA282548525 Enamine

2-(2-chlorophenyl)-N-(3-hydroxypropyl)acetamide

  • MDL number MFCD12178389
  • Cas Number : 1156897-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61195
Availability details for
Quantity
  • Availability
  • Add to Cart