ENA282548527 Enamine

N-[(furan-2-yl)methyl]-N-methyl-1,3-thiazolidine-4-carboxamide

  • MDL number MFCD12191552
  • Cas Number : 1218298-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61198
Availability details for
Quantity
  • Availability
  • Add to Cart