ENA282548528 Enamine

3-amino-1-(4-phenylpiperazin-1-yl)propan-1-one

  • MDL number MFCD06742065
  • Cas Number : 896522-90-0
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61199
Availability details for
Quantity
  • Availability
  • Add to Cart