ENA282548531 Enamine

2-[(4-aminophenyl)formamido]acetamide

  • MDL number MFCD09045995
  • Cas Number : 46319-19-1
  • Molecular Weight : 193.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60296
Availability details for
Quantity
  • Availability
  • Add to Cart