ENA282548533 Enamine

2-hydroxycyclopentane-1-carbonitrile

  • MDL number MFCD11632747
  • Cas Number : 57836-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61208

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61208
Availability details for
Quantity
  • Availability
  • Add to Cart