ENA282548536 Enamine

N,N-dimethyl-1H-indole-3-carboxamide

  • MDL number MFCD00983879
  • Cas Number : 27409-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234...236
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61217

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61217
Availability details for
Quantity
  • Availability
  • Add to Cart