ENA282548537 Enamine

N-methyl-1H-indole-3-carboxamide

  • MDL number MFCD12193321
  • Cas Number : 85729-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61218

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61218
Availability details for
Quantity
  • Availability
  • Add to Cart