ENA282548541 Enamine

N-cyclopentyl-4-hydroxybenzamide

  • MDL number MFCD11130714
  • Cas Number : 1019403-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61222

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61222
Availability details for
Quantity
  • Availability
  • Add to Cart