ENA282548551 Enamine

4-(3-methoxyphenyl)piperidine hydrochloride

  • MDL number MFCD02178922
  • Cas Number : 325808-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61234

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61234
Availability details for
Quantity
  • Availability
  • Add to Cart