ENA282548552 Enamine

1-(pyridine-3-carbonyl)piperazine dihydrochloride

  • MDL number MFCD08456890
  • Cas Number : 32422-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61235
Availability details for
Quantity
  • Availability
  • Add to Cart