ENA282548556 Enamine

7-bromo-5-chloro-3-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD09965715
  • Cas Number : 1019111-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61239
Availability details for
Quantity
  • Availability
  • Add to Cart