ENA282548558 Enamine

5-chloro-2-(trifluoromethyl)-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD11042742
  • Cas Number : 40851-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61241
Availability details for
Quantity
  • Availability
  • Add to Cart