ENA282548560 Enamine

[1-(thiophen-2-yl)cyclopentyl]methanamine

  • MDL number MFCD09694079
  • Cas Number : 75180-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61244

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61244
Availability details for
Quantity
  • Availability
  • Add to Cart