ENA282548562 Enamine

N-hexylpiperidine-3-carboxamide

  • MDL number MFCD12079153
  • Cas Number : 1153987-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61246
Availability details for
Quantity
  • Availability
  • Add to Cart