ENA282548563 Enamine

4-(cyclohexylsulfanyl)-2-methylaniline

  • MDL number MFCD12801138
  • Cas Number : 1178225-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61247

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61247
Availability details for
Quantity
  • Availability
  • Add to Cart