ENA282548565 Enamine

4-(butan-2-ylsulfanyl)-2-methylaniline

  • MDL number MFCD12169278
  • Cas Number : 1155084-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-61249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61249
Availability details for
Quantity
  • Availability
  • Add to Cart