ENA282548566 Enamine

N-(butan-2-yl)-2-methyl-1,2,3,4-tetrahydroquinoline-4-carboxamide

  • MDL number MFCD09932162
  • Cas Number : 1134943-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61250
Availability details for
Quantity
  • Availability
  • Add to Cart