ENA282548574 Enamine

2-{imidazo[1,2-a]pyridin-2-ylmethoxy}benzaldehyde

  • MDL number MFCD06316567
  • Cas Number : 631858-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123...125
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61267

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61267
Availability details for
Quantity
  • Availability
  • Add to Cart