ENA282548578 Enamine

5-cyclopropyl-1-(4-fluorophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD10003269
  • Cas Number : 1017423-32-3
  • Molecular Weight : 247.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61272
Availability details for
Quantity
  • Availability
  • Add to Cart